Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078166.1 | NP_001071634.1 | 1 | 307 | ||
NM_006924.4 | NP_008855.1 | 1 | 307 | |||
NR_034041.1 | 1 | 307 | ||||
GenBank mRNA | AB062124.1 | - | 1 | 188 | ||
AB209558.1 | - | 1 | 194 | |||
AK225711.1 | - | 1 | 185 | |||
AK289933.1 | - | 1 | 216 | |||
AK312781.1 | - | 1 | 204 | |||
BC010264.1 | - | 1 | 133 | 2900101 | ||
BC033785.1 | - | 1 | 193 | 5166554 | ||
DA939911.1 | - | 1 | 307 | |||
M69040.1 | - | 1 | 222 | |||
M72709.1 | - | 1 | 194 |