siRNA Details
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037940.4 | NP_001033029.1 | 7, 8 | 876 | ||
NM_001127367.1 | NP_001120839.1 | 7, 8 | 873 | |||
NM_011847.4 | NP_035977.2 | 7, 8 | 876 | |||
GenBank mRNA | AB028854.1 | - | 7, 8 | 778 | ||
AF035962.1 | - | 7, 8 | 776 | |||
AK032039.1 | - | 7, 8 | 785 | |||
AK133805.1 | - | 7, 8 | 793 | |||
AK134387.1 | - | 7, 8 | 835 | |||
AK144182.1 | - | 7, 8 | 797 | |||
AK151219.1 | - | 8, 9 | 783 | |||
AK151752.1 | - | 7, 8 | 750 | |||
AK152626.1 | - | 7, 8 | 790 | |||
AK152632.1 | - | 7, 8 | 750 | |||
AK153091.1 | - | 7, 8 | 756 | |||
AK169332.1 | - | 7, 8 | 782 | |||
AK169767.1 | - | 7, 8 | 744 | |||
BC003702.1 | - | 7, 8 | 758 | 3601332 | ||
BC083349.1 | - | 7, 8 | 768 | 30541723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
