Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF037205.1 | - | 8, 9 | 795 | ||
AF037206.1 | - | 8, 9 | 795 | |||
AK036591.1 | - | 8, 9 | 840 | |||
AK050095.1 | - | 8, 9 | 1540 | |||
AK139447.1 | - | 8, 9 | 801 | |||
AK140682.1 | - | 8, 9 | 851 | |||
AK158046.1 | - | 8, 9 | 836 | |||
AK167278.1 | - | 8, 9 | 800 | |||
BC058182.1 | - | 8, 9 | 803 | 5324040 |