Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167574.1 | NP_001161046.1 | 2 | 215 | ||
NM_006953.3 | NP_008884.1 | 2 | 215 | |||
GenBank mRNA | AB010116.1 | - | 1 | 92 | ||
AB010637.1 | - | 2 | 188 | |||
AF085808.1 | - | 2 | 220 | |||
BC069544.1 | - | 2 | 220 | 7262218 | ||
BC108900.2 | - | 2 | 211 | 40004236 | ||
BM727658.1 | - | 2 | 194 | |||
CR456608.1 | - | 2 | 190 |