Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011931.3 | NP_036061.1 | 11 | 1286 | ||
GenBank mRNA | AF151110.2 | - | 11 | 1174 | ||
BC082804.1 | - | 11 | 1286 | 6852010 | ||
BC158002.1 | - | 11 | 1194 | 9007311 |