Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK3 SIRNA Assays ›
          Gene Symbol:
          Mapk3
          Gene Name:
          mitogen-activated protein kinase 3
          Gene Aliases:
          Erk-1, Erk1, Ert2, Esrk1, Mnk1, Mtap2k, Prkm3, p44, p44erk1, p44mapk
          Chromosome Location:
          Chr.7: 126759626 - 126765816 on Build GRCm38
          UniGene ID:
          Mm.8385
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 188103

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mapk3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011952.2 NP_036082.1 1, 2 201
          GenBank mRNA AK155287.1 - 1 143
          BC013754.1 - 1, 2 34
          BC029712.1 - 1, 2 195 5041517
          BQ929318.1 - 1, 2 201
          Z14249.1 - 1, 2 172

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          MAPK import into nucleus
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          protein complex assembly
          protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          DNA damage induced protein phosphorylation
          cell cycle
          signal transduction
          response to toxic substance
          organ morphogenesis
          phosphorylation
          peptidyl-serine phosphorylation
          sensory perception of pain
          BMP signaling pathway
          positive regulation of cyclase activity
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          regulation of stress-activated MAPK cascade
          positive regulation of histone phosphorylation
          positive regulation of histone acetylation
          intracellular signal transduction
          peptidyl-tyrosine autophosphorylation
          response to exogenous dsRNA
          positive regulation of translation
          positive regulation of transcription from RNA polymerase II promoter
          regulation of sequence-specific DNA binding transcription factor activity
          cartilage development
          regulation of cytoskeleton organization
          lung morphogenesis
          trachea formation
          ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          interleukin-1-mediated signaling pathway
          response to epidermal growth factor
          caveolin-mediated endocytosis
          regulation of Golgi inheritance
          regulation of early endosome to late endosome transport
          negative regulation of apolipoprotein binding
          nucleotide binding
          phosphotyrosine binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphatase binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0