Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006861.6 | NP_006852.1 | 5 | 653 | ||
XM_011537779.1 | XP_011536081.1 | 5 | 590 | |||
GenBank mRNA | AF498960.1 | - | 5 | 464 | ||
AK125543.1 | - | 6 | 1428 | |||
AK312538.1 | - | 5 | 563 | |||
BC015931.2 | - | 5 | 568 | 3907209 | ||
BM467132.1 | - | 5 | 635 | |||
BQ233709.1 | - | 5 | 609 | |||
BT020024.1 | - | 5 | 464 | |||
CR536486.1 | - | 5 | 464 | |||
CR541683.1 | - | 5 | 464 | |||
X79781.1 | - | 5 | 545 |