Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083963.1 | NP_001077432.1 | 3 | 348 | ||
NM_001083964.1 | NP_001077433.1 | 3 | 373 | |||
NM_001083965.1 | NP_001077434.1 | 3 | 373 | |||
NM_006862.3 | NP_006853.2 | 3 | 373 | |||
XM_005244856.4 | XP_005244913.1 | 3 | 239 | |||
XM_017000122.1 | XP_016855611.1 | 3 | 415 | |||
XM_017000123.1 | XP_016855612.1 | 3 | 415 | |||
XM_017000124.1 | XP_016855613.1 | 3 | 314 | |||
XM_017000125.1 | XP_016855614.1 | 3 | 343 | |||
XM_017000126.1 | XP_016855615.1 | 3 | 314 | |||
XM_017000127.1 | XP_016855616.1 | 3 | 343 | |||
XM_017000128.1 | XP_016855617.1 | 3 | 368 | |||
XM_017000129.1 | XP_016855618.1 | 3 | 333 | |||
XM_017000130.1 | XP_016855619.1 | 3 | 307 | |||
GenBank mRNA | AF119121.2 | - | 3 | 361 | ||
AF227192.1 | - | 3 | 361 | |||
AK056402.1 | - | 3 | 308 | |||
AK225160.1 | - | 3 | 327 | |||
AK226092.1 | - | 3 | 286 | |||
AK295948.1 | - | 3 | 244 | |||
AK303063.1 | - | 3 | 269 | |||
AK316279.1 | - | 3 | 324 | |||
BC032690.1 | - | 3 | 280 | |||
DA796638.1 | - | 3 | 373 |