Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other XBP1 SIRNA Assays ›
          Gene Symbol:
          Xbp1
          Gene Name:
          X-box binding protein 1
          Gene Aliases:
          D11Ertd39e, TREB-5, TREB5, XBP-1
          Chromosome Location:
          Chr.11: 5520641 - 5525993 on Build GRCm38
          UniGene ID:
          Mm.469937
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 188594

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Xbp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271730.1 NP_001258659.1 6 1178
          NM_013842.3 NP_038870.2 5 1204
          GenBank mRNA AF027963.3 - 5 878
          AF443192.1 - 6 1178
          AK007724.1 - 5 862
          AK145241.1 - 5 865
          BC003496.1 - 1 334
          BC008153.1 - 5 819
          BC016079.1 - 5 1098 4489182
          BC029197.1 - 5 859 2648085
          BC035452.1 - 1 27

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          liver development
          positive regulation of protein phosphorylation
          endothelial cell proliferation
          epithelial cell maturation
          positive regulation of immunoglobulin production
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          ubiquitin-dependent protein catabolic process
          lipid metabolic process
          transport
          autophagy
          apoptotic process
          response to unfolded protein
          positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response
          multicellular organism development
          muscle organ development
          regulation of autophagy
          positive regulation of autophagy
          negative regulation of myotube differentiation
          phosphatidylinositol 3-kinase signaling
          protein transport
          cell growth
          cell differentiation
          endoplasmic reticulum unfolded protein response
          exocrine pancreas development
          positive regulation of histone methylation
          regulation of protein stability
          protein destabilization
          cellular response to nutrient
          positive regulation of TOR signaling
          cellular response to insulin stimulus
          cellular response to oxidative stress
          response to endoplasmic reticulum stress
          cellular triglyceride homeostasis
          cellular response to vascular endothelial growth factor stimulus
          cellular response to glucose starvation
          response to drug
          glucose homeostasis
          cholesterol homeostasis
          positive regulation of transcription factor import into nucleus
          negative regulation of apoptotic process
          positive regulation of MHC class II biosynthetic process
          positive regulation of B cell differentiation
          positive regulation of T cell differentiation
          positive regulation of fat cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          vascular endothelial growth factor receptor signaling pathway
          neuron development
          positive regulation of immunoglobulin secretion
          response to electrical stimulus
          fatty acid homeostasis
          sterol homeostasis
          serotonin secretion, neurotransmission
          adipose tissue development
          epithelial cell maturation involved in salivary gland development
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to lipopolysaccharide
          cellular response to amino acid stimulus
          cellular response to fructose stimulus
          cellular response to glucose stimulus
          cellular response to interleukin-4
          cellular response to peptide hormone stimulus
          cellular response to fluid shear stress
          cellular response to laminar fluid shear stress
          positive regulation of plasma cell differentiation
          negative regulation of endoplasmic reticulum unfolded protein response
          positive regulation of endoplasmic reticulum unfolded protein response
          positive regulation of proteasomal protein catabolic process
          positive regulation of protein acetylation
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of lactation
          response to insulin-like growth factor stimulus
          positive regulation of hepatocyte proliferation
          positive regulation of endothelial cell apoptotic process
          positive regulation of interleukin-6 secretion
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter binding
          RNA polymerase II transcription factor binding
          enhancer sequence-specific DNA binding
          protease binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein kinase binding
          chromatin DNA binding
          ubiquitin protein ligase binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          histone deacetylase binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0