Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CUL3 SIRNA Assays ›
Gene Symbol:
Cul3
Gene Name:
cullin 3
Gene Aliases:
AI467304, AW146203, mKIAA0617
Chromosome Location:
Chr.1: 80265610 - 80340762 on Build GRCm38
UniGene ID:
Mm.12665
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
188654
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Cul3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_016716.4
NP_057925.1
5
671
GenBank mRNA
AF129738.1
-
5
561
AK129174.1
-
6
1059
BC027304.1
-
5
696
3981170
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106923, XM_008767226, XM_008767227, XM_008767228, XM_008767229, and others.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
mitotic cell cycle
cell morphogenesis
in utero embryonic development
trophectodermal cellular morphogenesis
regulation of transcription from RNA polymerase II promoter
ubiquitin-dependent protein catabolic process
protein monoubiquitination
transport
ER to Golgi vesicle-mediated transport
mitotic metaphase plate congression
integrin-mediated signaling pathway
gastrulation
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
Wnt signaling pathway
vesicle-mediated transport
cell migration
protein ubiquitination
stem cell division
positive regulation of cytokinesis
negative regulation of Rho protein signal transduction
embryonic cleavage
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
fibroblast apoptotic process
positive regulation of mitotic metaphase/anaphase transition
COPII vesicle coating
liver morphogenesis
ubiquitin-protein transferase activity
protein binding
cyclin binding
POZ domain binding
ubiquitin protein ligase binding
protein homodimerization activity
protein heterodimerization activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-v2dhc:80/100.66.79.56:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0