Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033622.1 | NP_296371.1 | 1 | 414 | ||
GenBank mRNA | AF119383.1 | - | 1 | 414 | ||
AF352245.1 | - | 1 | 418 | |||
AK034121.1 | - | 1 | 46 | |||
AK050384.1 | - | 1 | 396 | |||
AK079180.1 | - | 1 | 575 | |||
AK165081.1 | - | 1 | 405 | |||
AY290823.1 | - | 1 | 198 | |||
BC106840.1 | - | 1 | 387 | 40045718 | ||
BC106841.1 | - | 1 | 387 | 40045720 |