Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012603.2 | NP_036735.2 | 2 | 719 | ||
GenBank mRNA | AY679729.1 | - | 2 | 556 | ||
AY679730.1 | - | 2 | 719 | |||
BC091699.1 | - | 2 | 525 | 7303974 |