Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017109.1 | NP_058805.1 | 5 | 801 | ||
GenBank mRNA | AF056704.1 | - | 5 | 801 |