Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MB SIRNA Assays ›
          Gene Symbol:
          Mb
          Gene Name:
          myoglobin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 118101633 - 118108864 on Build Rnor_6.0
          UniGene ID:
          Rn.40511
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 189190

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021588.2 NP_067599.1 3 601
          GenBank mRNA BC070511.1 - 3 601 7103504
          FQ214690.1 - 3 633
          FQ214898.1 - 3 632
          FQ214986.1 - 3 633
          FQ215026.1 - 3 633
          FQ215032.1 - 3 632
          FQ215043.1 - 3 632
          FQ215217.1 - 3 631
          FQ215441.1 - 3 632
          FQ215511.1 - 3 631
          FQ215553.1 - 3 632
          FQ215560.1 - 3 631
          FQ215564.1 - 3 631
          FQ215642.1 - 3 642
          FQ215802.1 - 3 633
          FQ215880.1 - 3 632
          FQ215982.1 - 3 632
          FQ216001.1 - 3 631
          FQ216029.1 - 3 630
          FQ216127.1 - 3 630
          FQ216280.1 - 3 639
          FQ216299.1 - 3 633
          FQ216306.1 - 3 634
          FQ216335.1 - 3 633
          FQ216380.1 - 3 632
          FQ216443.1 - 3 631
          FQ216504.1 - 3 632
          FQ216709.1 - 3 631
          FQ216731.1 - 3 631
          FQ216841.1 - 3 630
          FQ217450.1 - 3 632
          FQ217496.1 - 3 631
          FQ218027.1 - 3 632
          FQ223624.1 - 3 631
          FQ223672.1 - 3 628
          FQ223774.1 - 3 633
          FQ223938.1 - 3 633
          FQ224156.1 - 3 629
          FQ224523.1 - 3 632
          FQ224833.1 - 1 123
          FQ224856.1 - 3 632
          FQ224909.1 - 3 632
          FQ224921.1 - 3 629
          FQ224975.1 - 3 632

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          heart development
          response to hormone
          oxygen transport
          slow-twitch skeletal muscle fiber contraction
          response to hydrogen peroxide
          enucleate erythrocyte differentiation
          brown fat cell differentiation
          oxygen transporter activity
          iron ion binding
          oxygen binding
          heme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9s72j:80/100.66.73.65:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0