Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022399.2 | NP_071794.1 | 4, 5 | 563 | ||
GenBank mRNA | BC062395.1 | - | 4, 5 | 540 | 5623178 | |
BU671467.1 | - | 4, 5 | 563 | |||
D78308.1 | - | 4, 5 | 515 | |||
FQ210587.1 | - | 568 | ||||
FQ220096.1 | - | 4, 5 | 569 | |||
X53363.1 | - | 4, 5 | 535 |