Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
189379
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
4
499
GenBank mRNA
BC062028.1
-
3
468
5623881
DN931898.1
-
3
517
FQ209537.1
-
4
499
FQ209603.1
-
4
499
FQ209620.1
-
3
484
FQ209649.1
-
4
499
FQ209757.1
-
4
500
FQ210253.1
-
4
499
FQ210412.1
-
4
499
FQ211152.1
-
4
499
FQ211263.1
-
4
498
FQ212554.1
-
4
499
FQ212743.1
-
4
499
FQ212875.1
-
4
499
FQ212927.1
-
4
500
FQ213085.1
-
4
499
FQ213427.1
-
4
499
FQ213510.1
-
4
499
FQ213847.1
-
4
500
FQ214087.1
-
4
499
FQ214354.1
-
4
499
FQ214515.1
-
4
499
FQ214553.1
-
4
500
FQ214850.1
-
4
499
FQ214997.1
-
4
499
FQ215007.1
-
4
499
FQ215548.1
-
4
501
FQ215662.1
-
4
499
FQ215682.1
-
4
499
FQ215870.1
-
4
499
FQ216120.1
-
4
500
FQ216296.1
-
4
499
FQ216389.1
-
4
500
FQ216406.1
-
4
502
FQ216495.1
-
4
499
FQ216618.1
-
4
499
FQ216733.1
-
4
501
FQ216774.1
-
4
499
FQ216813.1
-
4
498
FQ216913.1
-
4
500
FQ218561.1
-
4
499
FQ218832.1
-
4
499
FQ218943.1
-
4
499
FQ219025.1
-
4
502
FQ219184.1
-
4
503
FQ219389.1
-
4
499
FQ219457.1
-
4
499
FQ219815.1
-
4
499
FQ219837.1
-
4
499
FQ219855.1
-
4
499
FQ219923.1
-
4
499
FQ220008.1
-
4
499
FQ220051.1
-
4
499
FQ220077.1
-
4
499
FQ220115.1
-
4
499
FQ220201.1
-
4
497
FQ220213.1
-
4
499
FQ220247.1
-
4
500
FQ220282.1
-
4
501
FQ220284.1
-
4
499
FQ220302.1
-
4
499
FQ220338.1
-
4
499
FQ220345.1
-
4
499
FQ220370.1
-
4
499
FQ220406.1
-
4
499
FQ220413.1
-
4
499
FQ220420.1
-
4
500
FQ220433.1
-
4
496
FQ220444.1
-
4
499
FQ220449.1
-
4
499
FQ220500.1
-
4
499
FQ220514.1
-
4
499
FQ220527.1
-
4
499
FQ220533.1
-
4
531
FQ220542.1
-
4
499
FQ220545.1
-
4
499
FQ220634.1
-
4
499
FQ220654.1
-
4
499
FQ220663.1
-
4
499
FQ220684.1
-
4
500
FQ220690.1
-
4
501
FQ220769.1
-
4
499
FQ220787.1
-
4
499
FQ220801.1
-
4
499
FQ220833.1
-
4
499
FQ220838.1
-
4
499
FQ220888.1
-
4
499
FQ220932.1
-
4
500
FQ221146.1
-
4
499
FQ221380.1
-
4
499
FQ221556.1
-
4
497
FQ222171.1
-
4
499
FQ222181.1
-
4
499
FQ222327.1
-
4
499
FQ222772.1
-
4
499
FQ222906.1
-
4
499
FQ223121.1
-
4
499
FQ223487.1
-
4
499
FQ224547.1
-
4
499
FQ224834.1
-
4
499
FQ224846.1
-
4
499
FQ225049.1
-
4
499
FQ225170.1
-
4
499
FQ225608.1
-
4
499
FQ225654.1
-
4
501
FQ225860.1
-
4
501
FQ225886.1
-
4
499
FQ226306.1
-
4
501
FQ227381.1
-
4
499
FQ227522.1
-
4
494
FQ227719.1
-
4
499
FQ227837.1
-
4
499
FQ227937.1
-
4
500
FQ227941.1
-
4
499
FQ227942.1
-
4
619
FQ228548.1
-
4
499
FQ228995.1
-
4
499
FQ228996.1
-
4
499
FQ229139.1
-
4
501
FQ229159.1
-
4
499
FQ229164.1
-
4
500
FQ229208.1
-
4
499
FQ229241.1
-
4
499
FQ229244.1
-
4
499
FQ229248.1
-
2
379
FQ229272.1
-
4
499
FQ229273.1
-
4
500
FQ229303.1
-
4
499
FQ229337.1
-
4
499
FQ229352.1
-
4
499
FQ229365.1
-
4
499
FQ229397.1
-
4
499
FQ229408.1
-
4
500
FQ229433.1
-
4
499
FQ229448.1
-
4
499
FQ229521.1
-
4
499
FQ229547.1
-
4
499
FQ229554.1
-
4
499
FQ229566.1
-
4
495
FQ229610.1
-
4
499
FQ229626.1
-
4
496
FQ229726.1
-
4
499
FQ229740.1
-
4
499
FQ229742.1
-
4
499
FQ229776.1
-
4
499
FQ229808.1
-
4
499
FQ229815.1
-
4
499
FQ229830.1
-
4
501
FQ229883.1
-
4
501
FQ229901.1
-
4
499
FQ229932.1
-
2
397
FQ229952.1
-
4
499
FQ229974.1
-
4
499
FQ230005.1
-
4
499
FQ230055.1
-
4
498
FQ230267.1
-
4
502
FQ230943.1
-
4
499
FQ231445.1
-
4
501
FQ231525.1
-
4
499
FQ231591.1
-
4
501
FQ231710.1
-
4
500
FQ231733.1
-
4
500
FQ231897.1
-
4
501
FQ231967.1
-
4
499
FQ232029.1
-
4
500
FQ232154.1
-
4
499
FQ232165.1
-
4
499
FQ232319.1
-
4
500
FQ232697.1
-
4
501
FQ232767.1
-
4
499
FQ232951.1
-
4
499
FQ233152.1
-
4
498
FQ233377.1
-
4
499
FQ233473.1
-
4
499
FQ233732.1
-
4
499
FQ233737.1
-
4
499
FQ233808.1
-
4
499
FQ234195.1
-
4
499
FQ234341.1
-
4
499
FQ234613.1
-
4
499
FQ234733.1
-
4
499
FQ234735.1
-
4
389
FQ235063.1
-
4
499
FQ235079.1
-
4
499
FQ235122.1
-
4
499
FQ235259.1
-
4
499
X15096.1
-
4
499
Z29530.1
-
3
455
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0