Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023981.4 | NP_076471.3 | 2 | 276 | ||
XM_006233187.1 | XP_006233249.1 | 2 | 147 | |||
XM_008761426.1 | XP_008759648.1 | 2 | 147 | |||
XM_008761427.1 | XP_008759649.1 | 2 | 147 | |||
XM_008761428.1 | XP_008759650.1 | 2 | 147 | |||
XM_008761429.1 | XP_008759651.1 | 2 | 147 | |||
GenBank mRNA | AF514355.1 | - | 2 | 116 | ||
AF514356.1 | - | 2 | 116 | |||
AF514357.1 | - | 2 | 126 | |||
AF515736.1 | - | 2 | 214 | |||
BC074007.1 | - | 2 | 276 | 7099454 |