Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053455.2 | NP_445907.2 | 1, 2 | 604 | ||
GenBank mRNA | AF323608.2 | - | 1, 2 | 631 | ||
BC072502.1 | - | 1, 2 | 553 | |||
BC085357.1 | - | 1, 2 | 561 | |||
BC107665.1 | - | 1, 2 | 577 |