Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057155.3 | NP_476496.1 | 3, 4 | 267 | ||
XM_008773513.1 | XP_008771735.1 | 4, 5 | 707 | |||
XM_008773514.1 | XP_008771736.1 | 3, 4 | 570 | |||
GenBank mRNA | AF359355.1 | - | 3, 4 | 267 | ||
BC074017.1 | - | 3, 4 | 531 | 7098467 |