Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VCP SIRNA Assays ›
          Gene Symbol:
          VCP
          Gene Name:
          valosin containing protein
          Gene Aliases:
          ALS14, CMT2Y, HEL-220, HEL-S-70, IBMPFD, IBMPFD1, TERA, p97
          Chromosome Location:
          Chr.9: 35056068 - 35072742 on Build GRCh38
          UniGene ID:
          Hs.529782
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 19081

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: VCP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007126.3 NP_009057.1 3 557
          GenBank mRNA AF100752.1 - 3 380
          AK307735.1 - 3 372
          AK312310.1 - 3 439
          BC110913.1 - 3 367 6502535
          BC121794.1 - 3 364 40108360
          BC122550.1 - 3 364 40108357
          DA880782.1 - 3 557
          FJ224344.1 - 3 380
          FJ224352.1 - 3 380

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          double-strand break repair
          NADH metabolic process
          ER to Golgi vesicle-mediated transport
          autophagy
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          positive regulation of mitochondrial membrane potential
          protein ubiquitination
          protein N-linked glycosylation via asparagine
          viral genome replication
          translesion synthesis
          ER-associated ubiquitin-dependent protein catabolic process
          endoplasmic reticulum unfolded protein response
          retrograde protein transport, ER to cytosol
          positive regulation of protein complex assembly
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein hexamerization
          ERAD pathway
          regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          establishment of protein localization
          positive regulation of protein catabolic process
          ATP metabolic process
          protein homooligomerization
          aggresome assembly
          error-free translesion synthesis
          ER-associated misfolded protein catabolic process
          flavin adenine dinucleotide catabolic process
          autophagosome maturation
          positive regulation of protein K63-linked deubiquitination
          positive regulation of Lys63-specific deubiquitinase activity
          regulation of aerobic respiration
          positive regulation of oxidative phosphorylation
          positive regulation of ubiquitin-specific protease activity
          positive regulation of ATP biosynthetic process
          protein binding
          ATP binding
          lipid binding
          ATPase activity
          protein phosphatase binding
          protein domain specific binding
          polyubiquitin binding
          ubiquitin protein ligase binding
          deubiquitinase activator activity
          K48-linked polyubiquitin binding
          MHC class I protein binding
          identical protein binding
          ADP binding
          ubiquitin-like protein ligase binding
          poly(A) RNA binding
          BAT3 complex binding
          ubiquitin-specific protease binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0