Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048180.1 | NP_001041645.1 | 9 | 2350 | ||
NM_134456.3 | NP_604451.2 | 9 | 2550 | |||
XM_006230351.2 | XP_006230413.1 | 8 | 4111 | |||
XM_006230352.2 | XP_006230414.1 | 8 | 4164 | |||
XM_006230353.2 | XP_006230415.1 | 9 | 2525 | |||
XM_006230354.2 | XP_006230416.1 | 9 | 3431 | |||
GenBank mRNA | AF047577.1 | - | 9 | 2350 | ||
U57391.1 | - | 9 | 2593 |