Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Potea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770390.1 | XP_008768612.1 | 1 | 338 | ||
XM_008770391.1 | XP_008768613.1 | 1 | 338 | |||
XM_008770392.1 | XP_008768614.1 | 1 | 338 | |||
GenBank mRNA | AB095021.1 | - | 1 | 207 | ||
BC083629.1 | - | 1 | 312 | 7134382 |