Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PICALM SIRNA Assays ›
          Gene Symbol:
          PICALM
          Gene Name:
          phosphatidylinositol binding clathrin assembly protein
          Gene Aliases:
          CALM, CLTH, LAP
          Chromosome Location:
          Chr.11: 85957171 - 86069881 on Build GRCh38
          UniGene ID:
          Hs.163893
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 19192

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PICALM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008660.2 NP_001008660.1 7 1022
          NM_001206946.1 NP_001193875.1 7 1022
          NM_001206947.1 NP_001193876.1 7 737
          NM_007166.3 NP_009097.2 7 1022
          XM_005274322.2 XP_005274379.1 7 1006
          XM_005274323.2 XP_005274380.1 7 1006
          XM_005274324.2 XP_005274381.1 7 1006
          XM_005274325.2 XP_005274382.1 7 1006
          XM_005274326.2 XP_005274383.1 7 1006
          XM_005274327.2 XP_005274384.1 7 1006
          XM_005274328.2 XP_005274385.1 7 1006
          XM_005274329.3 XP_005274386.1 7 1006
          XM_005274330.2 XP_005274387.1 7 1006
          XM_005274331.2 XP_005274388.1 7 1006
          XM_005274332.2 XP_005274389.1 7 1006
          XM_005274333.2 XP_005274390.1 7 1006
          XM_005274334.2 XP_005274391.1 7 1006
          XM_005274335.2 XP_005274392.1 7 1006
          XM_005274336.2 XP_005274393.1 7 1006
          XM_005274337.2 XP_005274394.1 7 1006
          XM_005274340.2 XP_005274397.1 7 1006
          XM_006718699.2 XP_006718762.1 7 1006
          XM_006718700.2 XP_006718763.1 7 1006
          XM_006718701.3 XP_006718764.1 7 1006
          XM_011545293.1 XP_011543595.1 7 1006
          XM_017018381.1 XP_016873870.1 7 1006
          XM_017018382.1 XP_016873871.1 7 1006
          XM_017018383.1 XP_016873872.1 7 1006
          XM_017018384.1 XP_016873873.1 7 1006
          XM_017018385.1 XP_016873874.1 7 1006
          XM_017018386.1 XP_016873875.1 7 1006
          XM_017018387.1 XP_016873876.1 7 1006
          XM_017018388.1 XP_016873877.1 7 1006
          GenBank mRNA AB210017.1 - 7 1022
          AB451308.1 - 7 705
          AK291414.1 - 8 889
          AK300275.1 - 7 714
          BC048259.1 - 7 783 5537605
          BC064357.1 - 7 984 5112878
          BC073961.1 - 7 858 6183858
          U45976.1 - 7 852

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053554, XM_006229676, XM_006229674, XM_006229681, XM_006229679, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001252521, NM_001252524, NM_001252523, NM_146194, NM_001252520, and others.

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          receptor-mediated endocytosis
          cell proliferation
          negative regulation of gene expression
          synaptic vesicle maturation
          vesicle-mediated transport
          endosomal transport
          regulation of endocytosis
          receptor internalization
          regulation of protein localization
          cargo loading into vesicle
          positive regulation of transcription, DNA-templated
          negative regulation of receptor-mediated endocytosis
          clathrin coat assembly
          iron ion homeostasis
          clathrin-mediated endocytosis
          iron ion import into cell
          cell-cell adhesion
          positive regulation of neuron death
          positive regulation of beta-amyloid formation
          regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process
          negative regulation of metalloendopeptidase activity involved in amyloid precursor protein catabolic process
          protein binding
          1-phosphatidylinositol binding
          clathrin binding
          clathrin heavy chain binding
          clathrin adaptor activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0