Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021868.2 | NP_068640.2 | 5 | 324 | ||
XM_006230622.2 | XP_006230684.1 | 5 | 350 | |||
XM_006230623.1 | XP_006230685.1 | 5 | 332 | |||
XM_006230625.2 | XP_006230687.1 | 5 | 350 | |||
XM_006230889.2 | XP_006230951.1 | 5 | 350 | |||
XM_006230890.1 | XP_006230952.1 | 5 | 332 | |||
XM_006230891.2 | XP_006230953.1 | 5 | 350 | |||
GenBank mRNA | AF054618.1 | - | 3 | 179 | ||
AF054619.1 | - | 3 | 179 | |||
BC081802.1 | - | 5 | 324 | 7107292 |