Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
Plec
Gene Name:
plectin
Gene Aliases:
Pcn, Plec1, Pltn
Chromosome Location:
Chr.7: 117230319 - 117291473 on Build Rnor_6.0
UniGene ID:
Rn.1085
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
192108
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Plec
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164296.1
NP_001157768.1
6, 7
704
NM_001164297.1
NP_001157769.1
6, 7
639
NM_001164298.1
NP_001157770.1
6, 7
698
NM_001164299.1
NP_001157771.1
6, 7
730
NM_001164302.1
NP_001157774.1
6, 7
684
NM_001164303.1
NP_001157775.1
6, 7
637
NM_001164304.1
NP_001157776.1
6, 7
897
NM_001164305.1
NP_001157777.1
6, 7
614
NM_001164307.1
NP_001157779.1
6, 7
711
NM_001164308.1
NP_001157780.1
6, 7
771
NM_022401.2
NP_071796.2
6, 7
1090
XM_006241870.2
XP_006241932.1
7, 8
1301
XM_006241875.2
XP_006241937.1
7, 8
1063
XM_006241882.2
XP_006241944.1
7, 8
1418
XM_008765592.1
XP_008763814.1
7, 8
1894
XM_008765593.1
XP_008763815.1
8, 9
1078
XM_008765594.1
XP_008763816.1
7, 8
2061
XM_008765595.1
XP_008763817.1
7, 8
2865
XM_008765596.1
XP_008763818.1
7, 8
688
XM_008765597.1
XP_008763819.1
7, 8
811
XM_008765598.1
XP_008763820.1
7, 8
1301
GenBank mRNA
AY480022.1
-
6, 7
704
AY480023.1
-
6, 7
639
AY480024.1
-
6, 7
698
AY480025.1
-
6, 7
730
AY480026.1
-
6, 7
684
AY480027.1
-
6, 7
1231
AY480028.1
-
6, 7
637
AY480029.1
-
6, 7
782
AY480030.1
-
6, 7
614
AY480031.1
-
6, 7
711
AY480032.1
-
6, 7
771
X59601.1
-
6, 7
1040
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
female pregnancy
response to nutrient
epithelial cell differentiation
hemidesmosome assembly
actin binding
structural constituent of cytoskeleton
cytoskeletal protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0