Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300791.1 | NP_001287720.1 | 7 | 1101 | ||
NM_001300792.1 | NP_001287721.1 | 7 | 1101 | |||
NM_007054.6 | NP_008985.3 | 7 | 1101 | |||
XM_006714526.3 | XP_006714589.1 | 7 | 1156 | |||
XM_017008995.1 | XP_016864484.1 | 7 | 1156 | |||
XM_017008996.1 | XP_016864485.1 | 7 | 1156 | |||
GenBank mRNA | AB209780.1 | - | 7 | 1049 | ||
AF041853.1 | - | 6 | 942 | |||
AK295089.1 | - | 7 | 1059 | |||
AK313359.1 | - | 7 | 1068 | |||
AM177178.1 | - | 6 | 942 | |||
BC020890.1 | - | 7 | 1066 | |||
BC032599.1 | - | 7 | 1084 | |||
BC039592.1 | - | 7 | 1058 | |||
BC045542.1 | - | 7 | 1095 | 5298675 |