Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMGA2 SIRNA Assays ›
Gene Symbol:
Hmga2
Gene Name:
high mobility group AT-hook 2
Gene Aliases:
Hmgic
Chromosome Location:
Chr.7: 65159944 - 65275408 on Build Rnor_6.0
UniGene ID:
Rn.29981
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
192639
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Hmga2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_032070.1
NP_114459.1
2
539
GenBank mRNA
U89695.1
-
2
539
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010441, X58380, BC052158, BC085085, BC069985, and others.
Panther Classification:
Molecular Function -
endodeoxyribonuclease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA catabolic process, endonucleolytic
epithelial to mesenchymal transition
chondrocyte differentiation
mesodermal-endodermal cell signaling
base-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
mitotic G2 DNA damage checkpoint
signal transduction
spermatogenesis
cell proliferation
positive regulation of cell proliferation
male gonad development
response to virus
regulation of cell cycle process
positive regulation of gene expression
cell proliferation in forebrain
pituitary gland development
adrenal gland development
chromosome breakage
heterochromatin assembly
negative regulation of intracellular steroid hormone receptor signaling pathway
somatic stem cell population maintenance
histone H2A-S139 phosphorylation
senescence-associated heterochromatin focus assembly
endodermal cell differentiation
chondrocyte proliferation
positive regulation of multicellular organism growth
DNA damage response, detection of DNA damage
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of DNA binding
negative regulation by host of viral transcription
fat cell differentiation
negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of JAK-STAT cascade
mesodermal cell differentiation
negative regulation of astrocyte differentiation
mesenchymal cell differentiation
stem cell differentiation
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of growth hormone secretion
adipose tissue development
fat pad development
positive regulation of cell cycle arrest
regulation of peptide hormone secretion
oncogene-induced cell senescence
regulation of stem cell population maintenance
positive regulation of stem cell proliferation
positive regulation of transcription regulatory region DNA binding
positive regulation of cellular response to X-ray
positive regulation of cellular senescence
positive regulation of response to DNA damage stimulus
negative regulation of double-strand break repair via nonhomologous end joining
regulation of cellular response to drug
regulatory region DNA binding
transcription factor activity, transcription factor binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
AT DNA binding
transcription factor activity, sequence-specific DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
DNA-dependent protein kinase activity
transcription factor binding
DNA binding, bending
enzyme binding
nucleosomal DNA binding
cAMP response element binding
MH2 domain binding
MH1 domain binding
SMAD binding
5'-deoxyribose-5-phosphate lyase activity
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0