Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POMT1 SIRNA Assays ›
Gene Symbol:
POMT1
Gene Name:
protein O-mannosyltransferase 1
Gene Aliases:
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
Chromosome Location:
Chr.9: 131502895 - 131523806 on Build GRCh38
UniGene ID:
Hs.522449
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19284
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077365.1
NP_001070833.1
6
662
NM_001077366.1
NP_001070834.1
5
510
NM_001136113.1
NP_001129585.1
6
902
NM_001136114.1
NP_001129586.1
4
403
NM_007171.3
NP_009102.3
6
662
XM_005272156.1
XP_005272213.1
6
788
XM_005272158.1
XP_005272215.1
5
517
XM_005272159.1
XP_005272216.1
4
407
XM_005272162.2
XP_005272219.1
6
636
XM_006716932.1
XP_006716995.1
5
440
XM_011518140.1
XP_011516442.1
5
515
XM_011518141.1
XP_011516443.1
5
515
XM_011518142.1
XP_011516444.1
4
317
XM_011518143.1
XP_011516445.1
4
408
XM_011518145.1
XP_011516447.1
3
183
XM_017014203.1
XP_016869692.1
6
1110
XM_017014204.1
XP_016869693.1
5
445
XM_017014205.1
XP_016869694.1
6
584
GenBank mRNA
AF095136.1
-
6
639
AK074874.1
-
6
620
AK074888.1
-
5
494
AK293529.1
-
4
315
AK295561.1
-
4
407
AK299646.1
-
3
214
AK300393.1
-
5
530
AK303254.1
-
3
315
AK307876.1
-
6
620
AK315218.1
-
6
597
BC022877.2
-
6
556
4826806
BC065268.1
-
6
847
6155142
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cell wall mannoprotein biosynthetic process
carbohydrate metabolic process
protein O-linked glycosylation
multicellular organism development
extracellular matrix organization
protein O-linked mannosylation
chain elongation of O-linked mannose residue
regulation of endoplasmic reticulum unfolded protein response
mannosyltransferase activity
dolichyl-phosphate-mannose-protein mannosyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0