Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007187.3 | NP_009118.1 | 3, 4 | 249 | ||
XM_005266245.2 | XP_005266302.1 | 3, 4 | 266 | |||
XM_017020380.1 | XP_016875869.1 | 3, 4 | 226 | |||
GenBank mRNA | AF071185.1 | - | 3, 4 | 285 | ||
AK000979.1 | - | 3, 4 | 301 | |||
AK001557.1 | - | 3, 4 | 284 | |||
AK314525.1 | - | 3, 4 | 286 | |||
BC104879.1 | - | 3, 4 | 193 | 8143882 | ||
BC108310.1 | - | 3, 4 | 250 | 6014541 |