Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN6 SIRNA Assays ›
Gene Symbol:
Ptpn6
Gene Name:
protein tyrosine phosphatase, non-receptor type 6
Gene Aliases:
Ptph6, Shp-1
Chromosome Location:
Chr.4: 157239141 - 157263890 on Build Rnor_6.0
UniGene ID:
Rn.18985
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
193032
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ptpn6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_053908.1
NP_446360.1
3
415
GenBank mRNA
U77038.1
-
3
415
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
negative regulation of humoral immune response mediated by circulating immunoglobulin
protein dephosphorylation
cell proliferation
positive regulation of cell proliferation
response to wounding
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cell differentiation
platelet formation
abortive mitotic cell cycle
positive regulation of cell adhesion mediated by integrin
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
megakaryocyte development
negative regulation of T cell proliferation
natural killer cell mediated cytotoxicity
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
regulation of B cell differentiation
negative regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of T cell receptor signaling pathway
regulation of release of sequestered calcium ion into cytosol
regulation of ERK1 and ERK2 cascade
platelet aggregation
regulation of G1/S transition of mitotic cell cycle
phosphotyrosine binding
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
SH3 domain binding
protein kinase binding
SH2 domain binding
natural killer cell lectin-like receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-n8jtk:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0