siRNA Details
Gene Symbol: Park7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277250.1 | NP_001264179.1 | 7 | 837 | ||
NM_001277251.1 | NP_001264180.1 | 8 | 896 | |||
NM_001277252.1 | NP_001264181.1 | 7 | 777 | |||
NM_001277253.1 | NP_001264182.1 | 7 | 889 | |||
NM_057143.2 | NP_476484.1 | 7 | 1019 | |||
GenBank mRNA | AF157511.1 | - | 7 | 1063 | ||
AF157512.1 | - | 7 | 817 | |||
AJ007291.1 | - | 8 | 944 | |||
FQ217993.1 | - | 3 | 466 | |||
FQ220895.1 | - | 7 | 889 | |||
FQ223564.1 | - | 7 | 887 | |||
FQ224480.1 | - | 8 | 880 |

More Information
Panther Classification:
Molecular Function -






