Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007257.5 | NP_009188.1 | 3 | 889 | ||
XM_011544365.2 | XP_011542667.1 | 3 | 988 | |||
GenBank mRNA | AB020690.1 | - | 1 | 296 | ||
AF037365.1 | - | 1 | 114 | |||
AK055325.1 | - | 3 | 831 | |||
BC036489.1 | - | 3 | 819 | 5262276 | ||
BC047515.1 | - | 3 | 794 | 5294654 | ||
BC062301.1 | - | 1 | 370 | 6580976 |