Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134370.2 | NP_599197.2 | 6 | 1784 | ||
XM_006231432.2 | XP_006231494.1 | 6 | 1700 | |||
XM_008760393.1 | XP_008758615.1 | 5 | 1696 | |||
XM_008760394.1 | XP_008758616.1 | 6 | 1889 | |||
XM_008760395.1 | XP_008758617.1 | 6 | 1891 | |||
XM_008760396.1 | XP_008758618.1 | 6 | 1680 | |||
GenBank mRNA | AB040468.1 | - | 3 | 775 |