Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AKR1C3 SIRNA Assays ›
          Gene Symbol:
          Akr1c3
          Gene Name:
          aldo-keto reductase family 1, member C3
          Gene Aliases:
          Akr1c18
          Chromosome Location:
          Chr.17: 69761126 - 69778021 on Build Rnor_6.0
          UniGene ID:
          Rn.10030
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 193560

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Akr1c3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_138510.1 NP_612519.1 1, 2 130
          GenBank mRNA BC086579.1 - 1, 2 129
          BC100248.1 - 1, 2 124 7307214
          D14424.1 - 1, 2 115
          L32601.1 - 1, 2 130

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          prostaglandin metabolic process
          progesterone catabolic process
          G-protein coupled receptor signaling pathway
          parturition
          response to nutrient
          steroid metabolic process
          positive regulation of cell proliferation
          male gonad development
          cellular response to starvation
          positive regulation of cell death
          farnesol catabolic process
          keratinocyte differentiation
          cellular response to reactive oxygen species
          progesterone metabolic process
          retinal metabolic process
          multicellular organismal macromolecule metabolic process
          daunorubicin metabolic process
          doxorubicin metabolic process
          regulation of retinoic acid receptor signaling pathway
          regulation of steroid biosynthetic process
          positive regulation of protein kinase B signaling
          oxidation-reduction process
          testosterone biosynthetic process
          cellular response to cadmium ion
          cellular response to calcium ion
          cellular response to prostaglandin stimulus
          cellular response to corticosteroid stimulus
          cellular response to jasmonic acid stimulus
          cellular response to prostaglandin D stimulus
          negative regulation of retinoic acid biosynthetic process
          regulation of testosterone biosynthetic process
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          retinal dehydrogenase activity
          alditol:NADP+ 1-oxidoreductase activity
          retinol dehydrogenase activity
          oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor
          phenanthrene 9,10-monooxygenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          geranylgeranyl reductase activity
          ketoreductase activity
          17-alpha,20-alpha-dihydroxypregn-4-en-3-one dehydrogenase activity
          15-hydroxyprostaglandin-D dehydrogenase (NADP+) activity
          androsterone dehydrogenase activity
          ketosteroid monooxygenase activity
          delta4-3-oxosteroid 5beta-reductase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cqs7j:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0