Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC102551418 SIRNA Assays ›
          Gene Symbol:
          LOC102551418 LOC102546456 LOC102547151 LOC102547206 LOC102547825 LOC102549424 LOC102551620 LOC102554594 LOC102555587 LOC102555695 LOC102555984 LOC103690189 LOC103693567
          Gene Name:
          uncharacterized LOC102551418
          uncharacterized LOC102546456
          uncharacterized LOC102547151
          uncharacterized LOC102547206
          uncharacterized LOC102547825
          uncharacterized LOC102549424
          uncharacterized LOC102551620
          uncharacterized LOC102554594
          uncharacterized LOC102555587
          uncharacterized LOC102555695
          uncharacterized LOC102555984
          uncharacterized LOC103690189
          MLV-related proviral Env polyprotein-like
          Gene Aliases:
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Chromosome Location:
          Chr.7: 77043608 - 77049143 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 168475038 - 168479805 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 153379326 - 153388347 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 50367844 - 50372609 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 109640649 - 109643104 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 45909951 - 45913620 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 88030140 - 88035306 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 12907304 - 12912069 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 162104142 - 162109315 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 41575520 - 41579189 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 162133656 - 162138397 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 60701661 - 60706426 on Build Rnor_6.0
          Chromosome Location:
          Chr.11: 45913549 - 45918378 on Build Rnor_6.0
          UniGene ID:
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 193749

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC102551418
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008765483.1 XP_008763705.1 2 759
          Gene Symbol: LOC102546456
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008764320.1 XP_008762542.1 1 230
          Gene Symbol: LOC102547151
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008761073.1 XP_008759295.1 4 412
          Gene Symbol: LOC102547206
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008763644.1 XP_008761866.1 1 230
          Gene Symbol: LOC102547825
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008769907.1 XP_008768129.1 1 200
          Gene Symbol: LOC102549424
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008768683.1 XP_008766905.1 1 206
          Gene Symbol: LOC102551620
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008762073.1 XP_008760295.1 1 524
          Gene Symbol: LOC102554594
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008758638.1 XP_008756860.1 1 230
          Gene Symbol: LOC102555587
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008764325.1 XP_008762547.1 2 407
          Gene Symbol: LOC102555695
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008768678.1 XP_008766900.1 1 206
          Gene Symbol: LOC102555984
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008764308.1 XP_008762530.1 1 205
          Gene Symbol: LOC103690189
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008766369.1 XP_008764591.1 1 230
          Gene Symbol: LOC103693567
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008768684.1 XP_008766906.1 3 2723

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0