siRNA Details
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153624.2 | NP_705888.2 | 25 | 4641 | ||
XM_006254403.2 | XP_006254465.1 | 25 | 4697 | |||
GenBank mRNA | AB086193.1 | - | 1 | 751 | ||
BC081716.1 | - | 2 | 933 | |||
BC166779.1 | - | 7 | 1681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
