Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173313.1 | NP_775435.1 | 3 | 382 | ||
GenBank mRNA | U25684.1 | - | 3 | 382 |