Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPX3 SIRNA Assays ›
          Gene Symbol:
          Gpx3
          Gene Name:
          glutathione peroxidase 3
          Gene Aliases:
          GPx-3, GPx-P, GSHPx-3, GSHPx-P, Gpxp
          Chromosome Location:
          Chr.10: 40247436 - 40255423 on Build Rnor_6.0
          UniGene ID:
          Rn.108074
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 194620

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gpx3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022525.3 NP_071970.2 4 511
          GenBank mRNA BC062227.1 - 4 487 6921653
          CK480346.1 - 4 511
          CV079715.1 - 3 250
          D00680.1 - 4 576
          FQ221037.1 - 4 483
          FQ221279.1 - 4 464
          FQ221454.1 - 4 483
          FQ223139.1 - 3 226
          FQ228485.1 - 4 483
          FQ228545.1 - 4 537
          FQ228679.1 - 4 483
          FQ228689.1 - 4 483
          FQ228690.1 - 4 496
          FQ228694.1 - 4 463
          FQ228697.1 - 4 483
          FQ228699.1 - 4 483
          FQ228705.1 - 4 481
          FQ228712.1 - 4 483
          FQ228719.1 - 4 483
          FQ228737.1 - 4 483
          FQ228751.1 - 4 483
          FQ228788.1 - 4 483
          FQ228802.1 - 4 483
          FQ228803.1 - 4 480
          FQ228804.1 - 4 483
          FQ228807.1 - 4 484
          FQ228808.1 - 4 483
          FQ228824.1 - 4 483
          FQ228834.1 - 4 485
          FQ228835.1 - 4 483
          FQ228845.1 - 4 483
          FQ228866.1 - 4 483
          FQ228878.1 - 4 483
          FQ228879.1 - 4 483
          FQ228886.1 - 4 483
          FQ228899.1 - 4 483
          FQ228911.1 - 4 483
          FQ228914.1 - 4 483
          FQ228921.1 - 4 483
          FQ228934.1 - 4 485
          FQ228945.1 - 4 483
          FQ228947.1 - 4 483
          FQ228960.1 - 4 483
          FQ228965.1 - 4 483
          FQ228968.1 - 4 483
          FQ228969.1 - 4 482
          FQ228970.1 - 4 483
          FQ228976.1 - 4 483
          FQ229118.1 - 4 483
          FQ229125.1 - 4 483
          FQ230052.1 - 3 226

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to molecule of fungal origin
          glutathione metabolic process
          response to oxidative stress
          female pregnancy
          response to organic substance
          response to selenium ion
          response to organic cyclic compound
          response to drug
          hydrogen peroxide catabolic process
          protein homotetramerization
          response to corticosterone
          oxidation-reduction process
          glutathione peroxidase activity
          selenium binding
          glutathione binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0