Search
Search

Gnb2| Gene Symbol: Gnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031037.2 | NP_112299.1 | 6 | 701 | ||
| GenBank mRNA | AF277892.1 | - | 5 | 482 | ||
| AF397193.1 | - | 2 | 281 | |||
| BC065579.1 | - | 6 | 701 | 5598923 | ||
| DQ120485.1 | - | 5 | 466 | |||
| DQ120486.1 | - | 5 | 466 | |||
| FQ209560.1 | - | 2 | 89 | |||
| U34959.1 | - | 5 | 473 | |||