Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other THRA SIRNA Assays ›
          Gene Symbol:
          Thra
          Gene Name:
          thyroid hormone receptor alpha
          Gene Aliases:
          ERBA1, Thra1, c-erbA-1
          Chromosome Location:
          Chr.10: 86657285 - 86684935 on Build Rnor_6.0
          UniGene ID:
          Rn.11307
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 195001

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Thra
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001017960.1 NP_001017960.1 3 517
          NM_031134.2 NP_112396.2 2 385
          GenBank mRNA CB580585.1 - 2 317
          M18028.1 - 3 407
          M31174.1 - 599
          X07409.1 - 1 46
          X12744.1 - 2 49

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          ossification
          kidney development
          liver development
          regulation of thyroid hormone mediated signaling pathway
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          brain development
          learning or memory
          regulation of heart contraction
          female courtship behavior
          response to cold
          hormone-mediated signaling pathway
          organ morphogenesis
          positive regulation of myotube differentiation
          negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
          erythrocyte differentiation
          lung development
          adrenal gland development
          intracellular receptor signaling pathway
          thyroid gland development
          response to nutrient levels
          regulation of myeloid cell apoptotic process
          response to drug
          cytoplasmic sequestering of transcription factor
          steroid hormone mediated signaling pathway
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of transcription, DNA-templated
          positive regulation of female receptivity
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          embryonic organ development
          regulation of lipid catabolic process
          Type I pneumocyte differentiation
          negative regulation of DNA-templated transcription, initiation
          steroid receptor RNA activator RNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          steroid hormone receptor activity
          single-stranded RNA binding
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          thyroid hormone receptor activity
          transcription factor binding
          zinc ion binding
          TBP-class protein binding
          protein domain specific binding
          chromatin DNA binding
          protein complex binding
          protein homodimerization activity
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          intronic transcription regulatory region DNA binding
          protein heterodimerization activity
          protein dimerization activity
          thyroid hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0