Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS12 SIRNA Assays ›
          Gene Symbol:
          Rps12 LOC100360573
          Gene Name:
          ribosomal protein S12
          ribosomal protein S12-like
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.1: 22758214 - 22760370 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 38619682 - 38620321 on Build Rnor_6.0
          UniGene ID:
          Rn.115713
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 195160

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031709.3 NP_113897.2 4, 5 351
          GenBank mRNA BC058460.1 - 4, 5 355 6919500
          FQ210032.1 - 4, 5 316
          FQ212402.1 - 4, 5 314
          FQ212636.1 - 4, 5 315
          FQ217474.1 - 4, 5 315
          FQ221068.1 - 4, 5 315
          FQ221108.1 - 4, 5 315
          FQ221199.1 - 4, 5 315
          FQ221249.1 - 4, 5 319
          FQ221263.1 - 4, 5 315
          FQ221353.1 - 4, 5 317
          FQ221368.1 - 4, 5 315
          FQ221420.1 - 4, 5 315
          FQ221550.1 - 4, 5 316
          FQ221711.1 - 4, 5 317
          FQ222180.1 - 4, 5 316
          FQ222260.1 - 4, 5 315
          FQ222326.1 - 4, 5 313
          FQ222422.1 - 4, 5 315
          FQ222486.1 - 4, 5 315
          FQ222589.1 - 4, 5 315
          FQ222591.1 - 4, 5 315
          FQ222620.1 - 4, 5 315
          FQ222621.1 - 4, 5 315
          FQ222742.1 - 4, 5 318
          FQ222863.1 - 4, 5 315
          FQ222888.1 - 4, 5 315
          FQ222895.1 - 4, 5 315
          FQ223029.1 - 4, 5 316
          FQ223134.1 - 4, 5 316
          FQ223216.1 - 4, 5 315
          FQ223305.1 - 4, 5 318
          FQ223374.1 - 4, 5 318
          FQ223466.1 - 4, 5 316
          FQ224074.1 - 4, 5 316
          FQ226180.1 - 4, 5 315
          FQ228598.1 - 4, 5 315
          FQ228992.1 - 4, 5 316
          M18547.1 - 4, 5 314
          Gene Symbol: LOC100360573
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002728597.4 XP_002728643.1 2 315

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_011295, AK166992, CJ207519, BC090257, AK008057, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          molecular_function
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0