Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LGALS3 SIRNA Assays ›
          Gene Symbol:
          Lgals3
          Gene Name:
          lectin, galactoside-binding, soluble, 3
          Gene Aliases:
          AGE-R3, CBP30, L-34, gal-3
          Chromosome Location:
          Chr.15: 24153602 - 24165537 on Build Rnor_6.0
          UniGene ID:
          Rn.764
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 195339

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lgals3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031832.1 NP_114020.1 5, 6 665
          GenBank mRNA BC089054.1 - 5, 6 653 7312891
          FQ210101.1 - 5, 6 675
          FQ214799.1 - 5, 6 947
          FQ218337.1 - 5, 6 665
          FQ219929.1 - 5, 6 665
          FQ219987.1 - 5, 6 665
          FQ220157.1 - 5, 6 665
          FQ220174.1 - 5, 6 664
          FQ220215.1 - 5, 6 667
          FQ220224.1 - 5, 6 659
          FQ220313.1 - 5, 6 665
          FQ220316.1 - 6, 7 895
          FQ220348.1 - 5, 6 665
          FQ220361.1 - 5, 6 824
          FQ220439.1 - 5, 6 664
          FQ220450.1 - 5, 6 665
          FQ220455.1 - 6, 7 892
          FQ220487.1 - 6, 7 865
          FQ220505.1 - 5, 6 665
          FQ220541.1 - 5, 6 665
          FQ220563.1 - 5, 6 665
          FQ220590.1 - 5, 6 667
          FQ220652.1 - 5, 6 665
          FQ220701.1 - 5, 6 664
          FQ220837.1 - 5, 6 664
          FQ220917.1 - 5, 6 665
          FQ220938.1 - 5, 6 665
          FQ220946.1 - 5, 6 665
          FQ220952.1 - 5, 6 665
          FQ221377.1 - 5, 6 665
          FQ223467.1 - 5, 6 667
          FQ223525.1 - 6, 7 457
          FQ223531.1 - 5, 6 665
          FQ229211.1 - 5, 6 669
          FQ229249.1 - 5, 6 665
          FQ229259.1 - 5, 6 665
          FQ229266.1 - 5, 6 665
          FQ229277.1 - 5, 6 662
          FQ229326.1 - 5, 6 665
          FQ229348.1 - 5, 6 665
          FQ229362.1 - 5, 6 665
          FQ229490.1 - 5, 6 665
          FQ229494.1 - 5, 6 665
          FQ229511.1 - 5, 6 665
          FQ229560.1 - 5, 6 667
          FQ229562.1 - 5, 6 667
          FQ229564.1 - 5, 6 667
          FQ229577.1 - 5, 6 665
          FQ229661.1 - 5, 6 665
          FQ229821.1 - 5, 6 665
          FQ229967.1 - 5, 6 667
          FQ229990.1 - 5, 6 667
          FQ229991.1 - 5, 6 667
          FQ229995.1 - 5, 6 665
          FQ230027.1 - 5, 6 665
          FQ230050.1 - 5, 6 668
          FQ230072.1 - 5, 6 665
          FQ230121.1 - 5, 6 672
          FQ230149.1 - 5, 6 667
          J02962.1 - 5, 6 665
          KF639951.1 - 4, 5 625
          KF639952.1 - 4, 5 625
          KF639953.1 - 4, 5 625
          KF639954.1 - 4, 5 625
          KF639955.1 - 5, 6 604
          KF639956.1 - 5, 6 604
          KF639957.1 - 5, 6 604
          KF639958.1 - 5, 6 604
          M13697.1 - 2, 3 253

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          extracellular matrix protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          monocyte chemotaxis
          mRNA processing
          signal transduction
          positive regulation of cell proliferation
          RNA splicing
          positive regulation of serotonin secretion
          extracellular matrix organization
          neutrophil chemotaxis
          epithelial cell differentiation
          regulation of T cell proliferation
          negative regulation of apoptotic process
          innate immune response
          positive regulation of angiogenesis
          negative regulation of endocytosis
          eosinophil chemotaxis
          macrophage chemotaxis
          negative regulation of T cell receptor signaling pathway
          positive chemotaxis
          regulation of T cell apoptotic process
          mononuclear cell migration
          positive regulation of mononuclear cell migration
          positive regulation of calcium ion import
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of cell proliferation in bone marrow
          negative regulation of immunological synapse formation
          negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          negative regulation of extrinsic apoptotic signaling pathway
          IgE binding
          Fc-gamma receptor I complex binding
          chemoattractant activity
          laminin binding
          poly(A) RNA binding
          monosaccharide binding
          disaccharide binding
          advanced glycation end-product receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-cqs7j:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0