Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053799.2 | NP_446251.1 | 7 | 533 | ||
XM_006249684.2 | XP_006249746.1 | 7 | 609 | |||
GenBank mRNA | BC072534.1 | - | 7 | 658 | 7098427 | |
J04487.1 | - | 7 | 534 |