Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAPK1 SIRNA Assays ›
          Gene Symbol:
          Mapk1
          Gene Name:
          mitogen activated protein kinase 1
          Gene Aliases:
          Erk2
          Chromosome Location:
          Chr.11: 88203785 - 88273288 on Build Rnor_6.0
          UniGene ID:
          Rn.34914
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 195780

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mapk1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053842.1 NP_446294.1 8 1152
          XM_006248658.2 XP_006248720.1 8 1189
          XM_006248659.2 XP_006248721.1 8 1189
          XM_008768848.1 XP_008767070.1 8 1189
          GenBank mRNA FQ211642.1 - 8 1192
          M64300.1 - 8 1152

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          MAPK import into nucleus
          protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          signal transduction
          positive regulation of cell proliferation
          response to toxic substance
          organ morphogenesis
          positive regulation of peptidyl-threonine phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          sensory perception of pain
          cytosine metabolic process
          positive regulation of cell migration
          regulation of protein stability
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          regulation of stress-activated MAPK cascade
          mammary gland epithelial cell proliferation
          intracellular signal transduction
          ERBB signaling pathway
          response to exogenous dsRNA
          response to estrogen
          negative regulation of cell differentiation
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          regulation of cytoskeleton organization
          long-term synaptic potentiation
          lung morphogenesis
          trachea formation
          labyrinthine layer blood vessel development
          ERK1 and ERK2 cascade
          response to epidermal growth factor
          cellular response to organic substance
          caveolin-mediated endocytosis
          regulation of Golgi inheritance
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          regulation of early endosome to late endosome transport
          phosphotyrosine binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase activity
          protein binding
          ATP binding
          transcription factor binding
          RNA polymerase II carboxy-terminal domain kinase activity
          kinase activity
          protein kinase binding
          phosphatase binding
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0