Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080583.1 | NP_542150.1 | 6 | 747 | ||
XM_008767935.1 | XP_008766157.1 | 7 | 1504 | |||
XM_008767936.1 | XP_008766158.1 | 6 | 718 | |||
XM_008767937.1 | XP_008766159.1 | 5 | 633 | |||
XM_008767938.1 | XP_008766160.1 | 5 | 650 | |||
XM_008767939.1 | XP_008766161.1 | 6 | 1486 | |||
GenBank mRNA | BC103481.1 | - | 7 | 716 | 7929959 | |
M34176.1 | - | 6 | 679 | |||
M77246.1 | - | 6 | 747 |
Gene Symbol: LOC103690090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773863.1 | XP_008772085.1 | 6 | 1486 | ||
XM_008773864.1 | XP_008772086.1 | 7 | 1504 | |||
XM_008773865.1 | XP_008772087.1 | 6 | 718 | |||
XM_008773866.1 | XP_008772088.1 | 5 | 633 | |||
XM_008773867.1 | XP_008772089.1 | 6 | 650 |