Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130779.2 | NP_570135.2 | 22 | 4588 | ||
XM_006239847.2 | XP_006239909.1 | 21 | 4324 | |||
XM_006239848.2 | XP_006239910.1 | 21 | 4327 | |||
XM_006239849.2 | XP_006239911.1 | 21 | 4321 | |||
XM_006239850.2 | XP_006239912.1 | 21 | 4327 | |||
XM_008764532.1 | XP_008762754.1 | 22 | 4444 | |||
GenBank mRNA | M55075.1 | - | 22 | 4510 |