Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153301.2 | NP_695213.1 | 10 | 1359 | ||
XM_008767779.1 | XP_008766001.1 | 9 | 1220 | |||
GenBank mRNA | AF415240.1 | - | 10 | 1367 |