Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19654
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
4
355
NM_012229.4
NP_036361.1
5
453
XM_005269632.4
XP_005269689.1
4
340
XM_005269633.4
XP_005269690.1
5
433
XM_005269634.4
XP_005269691.1
5
465
XM_005269635.4
XP_005269692.1
4
655
XM_005269636.4
XP_005269693.1
5
465
XM_005269637.4
XP_005269694.1
3
1159
XM_005269638.4
XP_005269695.1
3
2460
XM_005269639.4
XP_005269696.1
3
208
XM_005269640.4
XP_005269697.1
6
480
XM_005269641.4
XP_005269698.1
5
382
XM_005269642.4
XP_005269699.1
4
307
XM_005269643.4
XP_005269700.1
3
247
XM_005269644.4
XP_005269701.1
6
480
XM_005269645.4
XP_005269702.1
5
382
XM_005269646.4
XP_005269703.1
4
308
XM_006717721.3
XP_006717784.1
5
406
XM_011539537.2
XP_011537839.1
5
465
XM_017015947.1
XP_016871436.1
2
209
XM_017015948.1
XP_016871437.1
6
589
XM_017015949.1
XP_016871438.1
5
382
XM_017015950.1
XP_016871439.1
5
382
XM_017015951.1
XP_016871440.1
4
308
XM_017015952.1
XP_016871441.1
4
321
XM_017015953.1
XP_016871442.1
5
716
XM_017015954.1
XP_016871443.1
4
655
XM_017015955.1
XP_016871444.1
4
321
XM_017015956.1
XP_016871445.1
4
655
XM_017015957.1
XP_016871446.1
5
382
XM_017015958.1
XP_016871447.1
6
480
XM_017015959.1
XP_016871448.1
4
308
XM_017015960.1
XP_016871449.1
4
247
XM_017015961.1
XP_016871450.1
4
642
XM_017015962.1
XP_016871451.1
4
308
XM_017015963.1
XP_016871452.1
4
391
XM_017015964.1
XP_016871453.1
4
391
XM_017015965.1
XP_016871454.1
3
268
XM_017015966.1
XP_016871455.1
3
247
XM_017015968.1
XP_016871457.1
6
480
XM_017015969.1
XP_016871458.1
5
382
XM_017015970.1
XP_016871459.1
4
1217
XM_017015971.1
XP_016871460.1
3
261
XM_017015972.1
XP_016871461.1
5
382
XM_017015973.1
XP_016871462.1
5
382
XM_017015974.1
XP_016871463.1
5
406
XM_017015975.1
XP_016871464.1
3
247
XM_017015976.1
XP_016871465.1
4
345
GenBank mRNA
AK291667.1
-
4
321
AK295593.1
-
3
497
AK312517.1
-
4
321
BC001595.1
-
4
404
3543103
BX648078.1
-
5
462
D38524.1
-
4
343
DA879821.1
-
5
422
DB169780.1
-
4
357
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0