Search
Search

XRN2| Gene Symbol: XRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317960.1 | NP_001304889.1 | 4 | 633 | ||
| NM_012255.4 | NP_036387.2 | 4 | 473 | |||
| XM_017027722.1 | XP_016883211.1 | 4 | 628 | |||
| XM_017027723.1 | XP_016883212.1 | 4 | 402 | |||
| XM_017027724.1 | XP_016883213.1 | 4 | 628 | |||
| GenBank mRNA | AF152169.1 | - | 4 | 376 | ||
| AK001573.1 | - | 4 | 443 | |||
| AK302846.1 | - | 4 | 337 | |||
| AK303312.1 | - | 3 | 284 | |||
| AK314602.1 | - | 4 | 412 | |||
| AL136841.1 | - | 4 | 399 | |||
| AY382900.1 | - | 3 | 288 | |||
| CF994999.1 | - | 4 | 480 | |||