Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012096.2 | NP_036228.1 | 6 | 545 | ||
XM_011533583.2 | XP_011531885.1 | 7 | 598 | |||
GenBank mRNA | AB037849.2 | - | 6 | 398 | ||
AF169797.1 | - | 6 | 456 | |||
AF424738.1 | - | 6 | 454 | |||
AK299008.1 | - | 7 | 587 | |||
AK310716.1 | - | 6 | 493 | |||
BC028599.2 | - | 6 | 545 | 4829430 |